- Antibody type:Primary
- Antigen name:Syntaxin 13
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:101-200/274
- Form:Liquid
- Gene ID:65033
- Antigen synonyms:Syntaxin-13|Dm Syx13|Syntaxin13|Syx-13|Syx13|l301470|CG11278|STX12_RAT|ff16|Syntaxin 13|CT31479|Syx 13
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:31 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from rat Syntaxin 13
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Syntaxin 13 is an integral membrane protein that belongs to the t-SNARE family, a group of proteins involved in protein transport. Confocal immunofluoresence and electron microscopy studies have shown that syntaxin 13 is primarily localised to tubular early and recycling endosomes, where it colocalises with transferrin receptor, and it is also localised in endosomal vacuoles. Syntaxin 13 has been found to be expressed in all tissues, with higher levels of the protein found in brain, lung, spleen, thymus and testes. Immunoprecipitation studies show that syntaxin 13 complexes with beta-SNAP, VAMP2/3, and SNAP25. The binding of this complex to alpha-SNAP and NSF is terminated in the presence of ATP. These results suggest that syntaxin 13 is a SNARE protein which mediates the recycling protein flow through tubulo vesicular recycling endosomes.
Type: Primary
Antigen: Syntaxin 13
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/274
Host: Rabbit
Isotype: IgG
Reactivity: Mouse