Order Entry
Norway
ContactUsLinkComponent
HepG2 Whole Cell Cell Lysate (Adult Tumor)
HepG2 Whole Cell Cell Lysate (Adult Tumor)
Katalog # NOVUNBP1-42569
Supplier:  Novus Biologicals
CAS Number:  
undefined
HepG2 Whole Cell Cell Lysate (Adult Tumor)
Katalog # NOVUNBP1-42569
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    1 mg
  • Tested applications:
    WB, ImmunoPrecipitation

Specifications

About this item

HepG2 Whole Cell Cell Lysate (Adult Tumor).

  • Source: Human

Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.