PicoKine Human SP-D; ELISA Kit
Catalog # BSBTEK1171
Supplier: Boster Bio
CAS Number:
Specifications
- Pk:1 KIT
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Description:Human SP-D PicoKine; ELISA kit
- Tests per kit:96 wells
Specifications
About this item
Sandwich high sensitivity ELISA kit for quantitative detection of human SP-D.
- Sensitivity, detect single digit picogram level of target protein
- Specificity, thoroughly tested for cross-reactivity with family members
- Stability, guaranteed to remain stable for up to 12 months with proper storage
- Selection, 1000+ kits available, covering most common ELISA targets
- Service, on-demand technical support directly from ELISA experts
Detect Human Sp D/SFTPD with <20 pg/ml sensitivity. Format: 96-well plate with removable strips. Compatible samples: cell culture supernates, cell lysates, serum and plasma (heparin, EDTA). This is a TMB colorimetric sandwich ELISA kit with short assay time and fast experiment set up. Sp D/SFTPD tissue specificity: Expressed in lung, brain, pancreas and adipose tissue (mainly mature adipocytes). .
96wells/kit, with removable strips.