Order Entry
Norway
ContactUsLinkComponent
Anti-KLRG1 Golden Syrian Hamster Antibody (violetFluor® 450) [clone: 2F1]
Anti-KLRG1 Golden Syrian Hamster Antibody (violetFluor® 450) [clone: 2F1]
Supplier:  Tonbo Biosciences
Certificates
Anti-KLRG1 Golden Syrian Hamster Antibody (violetFluor® 450) [clone: 2F1]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 75-5893-U100
  • 75-5893-U025
  • Antigen symbol:
  • KLRG1
  • KLRG1
  • Antigen name:
  • Killer cell lectin-like receptor subfamily G member 1
  • Killer cell lectin-like receptor subfamily G member 1
  • Antigen synonyms:
  • MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
  • MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
  • Conjugation:
  • VioletFluor® 450
  • VioletFluor® 450
  • Clone:
  • 2F1
  • 2F1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Golden syrian hamster
  • Golden syrian hamster
  • Gene ID:
  • 50928
  • 50928
  • Isotype:
  • IgG
  • IgG
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications