Tin(II) chloride dihydrate USP
:10025-69-1
:
:SnCl₂·2H₂O
:MFCD00149863
:225.65 g/mol
:623 °C (1013 hPa)
:246 °C
:2.71 g/cm³ (25 °C)
:Ambient
:231-868-0
:3260
:8,III
:GHS05
:GHS07
- IDENTIFICATION (A):BLACKISH-GREY PRECIPITATE FORMS
- IDENTIFICATION (B):YELLOWISH,FLOCCULENT PRECIPITATE IS FORMED
- ANALYSIS 1::A YELLOWISH, FLOCCULENT PRICIPITATE FORMED
- ANALYSIS 2::PRECIPITATE DISSOLVES, SOLUTION IS CLEAR AND COLORLESS
- IDENTIFICATION (C):POSITIVE FOR CHLORIDE
- ASSAY (SnCl2•2H2O):98.0 - 102.0 %
- SULFATE:Max. 500 ppm
- IRON:Max. 100 ppm
- LEAD (Pb):Max. 50 µg/g
- APPEARANCE OF SOLUTION:CLEAR AND COLORLESS
- SUBSTANCES NOT PRECIPITATED BY THIOACETAMIDE:Max. 0.2 %
- ELEMENTAL IMPURITIES:AS REPORTED
- RESIDUAL SOLVENTS:AS REPORTED