Calcium chloride ≥98.0% (dried basis), powder, AnalaR NORMAPUR® Reag. Ph. Eur.
:10043-52-4
:
:CaCl₂
:MFCD00010903
:14,01659
:1600 °C (1013 hPa)
:772 °C
:2.174 g/cm³ (20 °C)
:110.98 g/mol
:Ambient
:233-140-8
:01-2119494219-28
:GHS07
- Assay (calculated on dried substance):≥ 98,0 %
- Free alkali (as Ca(OH)₂):≤ 0,2 %
- Loss on drying (200°C):≤ 5,0 %
- Water-absorption capacity(24 h;100 % RH):≥ 25,0 %
- Conforms to Reag. Ph.Eur.:Passes test