Cold resistant gloves, EDGE® 48-193
Supplier: Ansell
Certificates
About this item
Cold-resistant safety gloves, offering mechanical protection for heavy-duty applications.
- PVC coating repels water, oils and greases, besides providing good dexterity and pliability at low temperatures
- Provides a barrier against cold, oils, greases and abrasions
- Durable polyester liner keeps hands warm and dry
Recommended for: Outdoor winter use, maintenance, storing and transportation.
Certifications: EN 388:2016 (3121A), EN 511 (011); Cat. II
Specifications
- Catalog No:
- ANSE48-193/10
- ANSE48-193/11
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Liner material:
- Polyester
- Polyester
- Finish:
- Fully coated, PVC bumpers
- Fully coated, PVC bumpers
- Cuff style:
- Safety cuff
- Safety cuff
- EN norm:
- EN 388:2016 (3121A), EN 511 (011)
- EN 388:2016 (3121A), EN 511 (011)
- Sterility:
- Non sterile
- Non sterile