Industrial gloves, EDGE® 48-128
: Ansell
Multipurpose, abrasion-resistant, nitrile-coated safety gloves offering oil resistance.
- Very light and comfortable
- High level of dexterity and flexibility
- Good level of abrasion and oil resistance
- Dark glove shows dirt contamination less quickly
- FDA-compliant materials: Gloves are safe for food packaging
These flexible high-dexterity work gloves feature a lightweight, seamless knitted dark grey polyester liner and elasticized knitwrist dipped into black straight nitrile coating. Thanks to their nitrile coating, EDGE® 48-128 protective gloves grant better abrasion and oil resistance, grip and cushioning than light and mediumweight knitted cotton gloves.
Recommended for: Assembly of small parts; general handling; packaging and quality control.
: EN 388:2016 (4121A); Cat. II
- Catalog No:
- ANSE48-128/9
- ANSE48-128/10
- ANSE48-128/11
- ANSE48-128/6
- ANSE48-128/7
- ANSE48-128/8
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Finish:
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Palm nitrile coated
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- EN 388:2016 (4121A)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile