


Silver nitrate ≥99.8%, crystals USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Merck Index:12,08661
Boiling Pt:444 °C (1013 hPa)
Melting Pt:210 °C
Density:4.35 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-853-9
UN:1493
ADR:5.1,II
REACH:01-2119513705-43
:GHS03
:GHS05
:GHS09
Specification Test Results
- Assay:99.8% Min.
Specification Test Results
Frequently Bought Together