Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium chloride
Ammonium chloride
Ammonium chloride
Ammonium chloride
Supplier:
Spectrum Chemical
Certificates
Expand image
Ammonium chloride
Supplier:
Spectrum Chemical
Certificates
About this item
CAS Number:
12125-02-9
:
Formula:
NH₄Cl
MDL Number:
MFCD00011420
:
53.49 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Documentation