Order Entry
Norway
Orders LinkContactUsLinkComponent
Papain
Papain
Supplier:  Spectrum Chemical
Certificates

2 Options Available Below

Product Details & Documents

Papain, powder, purified, is a cysteine protease enzyme the is present in the papaya fruit.

It is traditionally used to break down meat fibres. It has more recently been used to dissociate cells in the early steps of cell culture preparation.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SPCMPA125-100GM
  • SPCMPA125-500GM
  • Enzyme name:
  • Papain
  • Papain
  • Form:
  • Powder, purified
  • Powder, purified
  • Purity:
  • Purified
  • Purified

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Poly bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - kr 1 950,00
    kr 1 950,00
    1 * 100 g
       kr 1 950,00

    Specifications:
    More
    Specifications
    Cat. No.
    SPCMPA125-100GM
    Enzyme name
    Papain
    Form
    Powder, purified
    Purity
    Purified
  • Poly bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - kr 5 675,00
    kr 5 675,00
    1 * 500 g
       kr 5 675,00

    Specifications:
    More
    Specifications
    Cat. No.
    SPCMPA125-500GM
    Enzyme name
    Papain
    Form
    Powder, purified
    Purity
    Purified