Tetrachloroethylene, anhydrous, Riedel-de Haën™
Catalog # HONC66287-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Bottle, septum-sealed
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:165.83 g/mol
Boiling Pt:120 - 122 °C (1,013 hPa)
Boiling Pt:120…122 °C (1,013 hPa)
Melting Pt:-22 °C
Melting Pt:–22 °C
Density:1.622…1.624 g/cm3 (20 °C)
Storage Temperature:Ambient
UN:1897
ADR:6.1,III
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834