Methyl acetate Analytical standard, reference material, Fluka™
Catalog # HONCR1920-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:79-20-9
- Description:Methyl acetate
- Storage conditions:RT
- MDL No.:MFCD00008711
- UN No.:1231
- Synonym(s):Acetic acid methyl ester
- Melting point:-98 °C
- Formula:C₃H₆O₂
- Molecular weight:74.08 g/mol
- Description:Methyl acetate
- Flash point:-12.99 °C
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery