Ethyl butyrate Analytical standard, reference material, Fluka™
Katalog # HONCR1904-1G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:1 g
- Standards:Analytical standard
- CAS No.:105-54-4
- Description:Ethyl butyrate
- Storage conditions:RT
- MDL No.:MFCD00009394
- UN No.:1180
- Synonym(s):Butyric acid ethyl ester
- Melting point:-11.4 °C
- Formula:C₆H₁₂O₂
- Molecular weight:116.16 g/mol
- Description:Ethyl butyrate
- Flash point:28 °C.972.3 hPa
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery