Monocrotophos Analytical standard, reference material, Fluka™
Katalog # HONCR1897-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:6923-22-4
- Description:Monocrotophos
- Storage conditions:2-8°C
- MDL No.:MFCD00055291
- UN No.:2811
- Synonym(s):Dimethyl (1E)-1-methyl-3-(methylamino)-3-oxo-1-propenyl phosphate
- Melting point:53 - 55 °C
- Formula:C₇H₁₄NO₅P
- Molecular weight:223.16 g/mol
- Description:Monocrotophos
- Physical form:Solid
Specifications
About this item
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery