Order Entry
Norway
ContactUsLinkComponent
Bromophos-methyl Analytical standard, reference material, Fluka™
Bromophos-methyl Analytical standard, reference material, Fluka™
  HONCR1888-100MG
 :  
Bromophos-methyl Analytical standard, reference material, Fluka™
  HONCR1888-100MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    2104-96-3
  • Description:
    Bromophos-methyl
  • Storage conditions:
    2-8°C
  • MDL No.:
    MFCD00055261
  • Synonym(s):
    O-(4-Bromo-2,5-dichlorophenyl) O,O-dimethyl phosphorothioate
  • Melting point:
    53 - 55 °C
  • Formula:
    C₈H₈BrCl₂O₃PS
  • Molecular weight:
    366.00 g/mol
  • Description:
    Bromophos-methyl
  • Physical form:
    Solid

 

 

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery