Cefalonium hydrate Analytical standard, reference material, Fluka™
Cefalonium hydrate, Analytical standard, Glass Bottle, 100 mg
Catalog # HONCR1872-100MG
Supplier: Honeywell Chemicals
Cefalonium hydrate Analytical standard, reference material, Fluka™
Catalog # HONCR1872-100MG
Supplier: Honeywell Chemicals
CAS Number:
Jump to:
Product Details & Documents
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard
- Description:Cefalonium hydrate
- Cat. No.:HONCR1872-100MG
- Formula:C₂₀H₁₈N₄O₅S₂·xH₂O
- Description:Cefalonium hydrate
- Storage conditions:2-8°C
- Physical form:Crystalline
- MDL No.:MFCD11975221
- Synonym(s):(6R,7R)-3-(4-(4-Carbamoyl-1-pyridiniomethyl)-8-oxo-7-(2-(2-thienyl)acetamido)-5-thia-1-azabicyclo(4.2.0)oct-2-en-2-carboxylate
- Molecular weight:458.51 g/mol
- CAS No.:5575-21-3



