Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Piperazine, Reference Standards

Supplier:   USP
Piperazine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1541907.
  • 1541601.
  • Description:
  • Piperazine Dihydrochloride
  • Piperazine
  • CAS No.:
  • 207605-49-0
  • 110-85-0
  • UN No.:
  • 2925

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1541907.
    Piperazine Dihydrochloride
    Piperazine Dihydrochloride
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - kr 3 000,00
    kr 3 000,00
    1 * 200 mg
       kr 3 000,00

    Specifications:
    More
    Specifications
    Cat. No.
    1541907.
    Description
    Piperazine Dihydrochloride
    CAS No.
    207605-49-0
    UN No.
  • 1541601.
    Piperazine
    Piperazine
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - kr 4 125,00
    kr 4 125,00
    1 * 200 mg
       kr 4 125,00

    Specifications:
    More
    Specifications
    Cat. No.
    1541601.
    Description
    Piperazine
    CAS No.
    110-85-0
    UN No.
    2925