Melengestrol Acetate, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1379254.
- 1379265.
- 1379276.
- Description:
- Melengestrol Acetate
- Melengestrol Acetate Related Compound A
- Melengestrol Acetate Related Compound B
- Synonym(s):
- 16-Methylene-3,20-dioxopregn-4-en-17-yl acetate
- 6,16-Dimethylene-3,20-dioxopregn-4-en-17-yl acetate
- CAS No.:
- 2919-66-6
- 6815-51-6
- 80097-73-0
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1379254.Melengestrol AcetateSpecifications:
More
Specifications
Cat. No.1379254.DescriptionMelengestrol AcetateSynonym(s)CAS No.2919-66-6 - 1379265.Melengestrol Acetate Related Compound AMelengestrol Acetate Related Compound AUSP reference standardVial25 mgLog in for availability1 * 25 mgkr 15 400,00Specifications:
More
Specifications
Cat. No.1379265.DescriptionMelengestrol Acetate Related Compound ASynonym(s)16-Methylene-3,20-dioxopregn-4-en-17-yl acetateCAS No.6815-51-6 - 1379276.Melengestrol Acetate Related Compound BMelengestrol Acetate Related Compound BUSP reference standardVial25 mgLog in for availability1 * 25 mgkr 14 025,00Specifications:
More
Specifications
Cat. No.1379276.DescriptionMelengestrol Acetate Related Compound BSynonym(s)6,16-Dimethylene-3,20-dioxopregn-4-en-17-yl acetateCAS No.80097-73-0
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



