Order Entry
Norway
Orders LinkContactUsLinkComponent
 

Felbamate, Reference Standards

 :   USP
Felbamate, Reference Standards
 :  USP
 
Felbamate, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1269312.
  • 1269323.
  • 1269334.
  • Description:
  • Felbamate
  • Felbamate Related Compound A
  • Felbamate Related Compound B
  • Synonym(s):
  • 3-Hydroxy-2-phenylpropyl carbamate
  • Phenethyl carbamate
  • CAS No.:
  • 25451-15-4
  • 25451-53-0
  • 6326-19-8

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Felbamate
    USP reference standard
    Vial
    250 mg
       
       
    1 * 250 mg
    1 * 250 mg - kr 13 600,00
    kr 13 600,00
    1 * 250 mg
       kr 13 600,00

     
     
    Specifications
    Cat. No.
    1269312.
    Description
    Felbamate
    Synonym(s)
    CAS No.
    25451-15-4
  • Felbamate Related Compound A
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - kr 14 300,00
    kr 14 300,00
    1 * 25 mg
       kr 14 300,00

     
     
    Specifications
    Cat. No.
    1269323.
    Description
    Felbamate Related Compound A
    Synonym(s)
    3-Hydroxy-2-phenylpropyl carbamate
    CAS No.
    25451-53-0
  • Felbamate Related Compound B
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - kr 12 475,00
    kr 12 475,00
    1 * 25 mg
       kr 12 475,00

     
     
    Specifications
    Cat. No.
    1269334.
    Description
    Felbamate Related Compound B
    Synonym(s)
    Phenethyl carbamate
    CAS No.
    6326-19-8