Order Entry
Norway
Orders LinkContactUsLinkComponent
Azacitidine, Reference Standards

Azacitidine, Reference Standards

Supplier:   USP
Azacitidine, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1045520.
  • 1045539.
  • 1045541.
  • 1045574.
  • Description:
  • Azacitidine
  • Azacitidine Related Compound A
  • Azacitidine Related Compound B
  • Azacitidine Related Compound C
  • Synonym(s):
  • 4-Amino-1,3,5-triazin-2(1H)-one
  • 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose
  • 1-β-D-Ribofuranosyl-3-guanylurea picrate
  • CAS No.:
  • 320-67-2
  • 931-86-2
  • 6974-32-9
  • 4336-46-3
  • Formula:
  • C₈H₁₂N₄O₅
  • C₇H₁₄N₄O₅C₆H₃N₃O₇
  • UN No.:
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1045520.
    Azacitidine
    Azacitidine
    USP reference standard
    Vial
    150 mg
    Log in for availability
    Log in for availability
    1 * 150 mg
    1 * 150 mg - kr 4 500,00
    kr 4 500,00
    1 * 150 mg
       kr 4 500,00

    Specifications:
    More
    Specifications
    Cat. No.
    1045520.
    Description
    Azacitidine
    Synonym(s)
    CAS No.
    320-67-2
    Formula
    C₈H₁₂N₄O₅
    UN No.
    3077
  • 1045539.
    Azacitidine Related Compound A
    Azacitidine Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - kr 14 650,00
    kr 14 650,00
    1 * 25 mg
       kr 14 650,00

    Specifications:
    More
    Specifications
    Cat. No.
    1045539.
    Description
    Azacitidine Related Compound A
    Synonym(s)
    4-Amino-1,3,5-triazin-2(1H)-one
    CAS No.
    931-86-2
    Formula
    UN No.
  • 1045541.
    Azacitidine Related Compound B
    Azacitidine Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - kr 13 600,00
    kr 13 600,00
    1 * 15 mg
       kr 13 600,00

    Specifications:
    More
    Specifications
    Cat. No.
    1045541.
    Description
    Azacitidine Related Compound B
    Synonym(s)
    1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose
    CAS No.
    6974-32-9
    Formula
    UN No.
  • 1045574.
    Azacitidine Related Compound C
    Azacitidine Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - kr 14 800,00
    kr 14 800,00
    1 * 15 mg
       kr 14 800,00

    Specifications:
    More
    Specifications
    Cat. No.
    1045574.
    Description
    Azacitidine Related Compound C
    Synonym(s)
    1-β-D-Ribofuranosyl-3-guanylurea picrate
    CAS No.
    4336-46-3
    Formula
    C₇H₁₄N₄O₅C₆H₃N₃O₇
    UN No.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.