Amitraz, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
: Cold Shipment required
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
: Cold Shipment required
- Catalog No:
- 1028019.
- 1028028.
- 1028032.
- Description:
- Amitraz Related Compound A
- Amitraz Related Compound B
- Amitraz Related Compound C
- Synonym(s):
- N-(2,4-Dimethylphenyl)formamide
- N'-(2,4-Dimethylphenyl)-N-methylformimidamide
- N,N'-Bis(2,4-dimethylphenyl)formimidamide
- CAS No.:
- 60397-77-5
- 33089-74-6
- 16596-04-6
- UN No.:
- 2811
Product Family Options
- DescriptionStandardsPack typePk
- Amitraz Related Compound AUSP reference standardVial40 mg1 * 40 mgkr 5 675,00
Specifications
Cat. No.1028019.DescriptionAmitraz Related Compound ASynonym(s)N-(2,4-Dimethylphenyl)formamideCAS No.60397-77-5UN No. - Amitraz Related Compound BUSP reference standardVial40 mg1 * 40 mgkr 7 575,00
Specifications
Cat. No.1028028.DescriptionAmitraz Related Compound BSynonym(s)N'-(2,4-Dimethylphenyl)-N-methylformimidamideCAS No.33089-74-6UN No.2811 - Amitraz Related Compound CUSP reference standardVial40 mg1 * 40 mgkr 5 850,00
Specifications
Cat. No.1028032.DescriptionAmitraz Related Compound CSynonym(s)N,N'-Bis(2,4-dimethylphenyl)formimidamideCAS No.16596-04-6UN No.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



