Metamizole sodium monohydrate
Catalog # ACRO467482500
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pakketype:Glass bottle
- Fp:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to light yellow to light beige powder or crystals
- Infrared spectrum:Conforms
- Melting point:211°C to 216°C
- HPLC:≥ 97.5%
- Water:4.5 to 5.5%
Specification Test Results
About this item
:
:351.36 g/mol
Merck Index:13.3385
:GHS08
Storage Temperature:Romtemperatur
CAS Number:5907-38-0
Formula:C₁₃H₁₆N₃NaO₄S·H₂O
MDL Number:MFCD00150293