Anti-TBK1 mouse monoclonal antibody [clone: 4E6]
Catalog # CAYM25924-100
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:TBK1
- Clonality:Monoclonal
- Clone:4E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Form:Liquid
- UniProtKB:Q9UHD2
- Storage buffer:PBS, pH 7,2, with 50% glycerol and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:Protein G-purified monoclonal antibody
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TBK1
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human