Order Entry
Norway
ContactUsLinkComponent
Anti-TNFRSF18 Rat Monoclonal Antibody [clone: DTA-1]
Anti-TNFRSF18 Rat Monoclonal Antibody [clone: DTA-1]
Catalog # 40-5874-U500
Supplier:  Tonbo Biosciences
undefined
Anti-TNFRSF18 Rat Monoclonal Antibody [clone: DTA-1]
Catalog # 40-5874-U500
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor Necrosis Factor Receptor Superfamily Member 18
  • Antigen symbol:
    TNFRSF18
  • Clone:
    DTA-1
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Neutralisation:
    Yes
  • Gene ID:
    21936
  • Antigen synonyms:
    CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
  • Storage temperature:
    Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
    Functional Assay
  • Size:
    500 µg
  • Pk:
    500 µG

Specifications

About this item

The DTA-1 antibody reacts with mouse CD357, also known as GITR or AITR (in humans), a 66-70 kDa member of the Tumor Necrosis Factor superfamily (TNFRSF18). GITR is primarily found on T cells, and its function may vary depending on the T cell type where it is expressed. GITR is upregulated on activated T cells where it provides co-stimulation, yet GITR may promote the inhibition of CD4+ CD25+ Treg cells, where it is expressed at high levels. GITR ligand (GITRL) is found on B cells, macrophages, dendritic and endothelial cells, and is implicated in regulating both innate and adaptive immune responses.

The DTA-1 antibody may be used for analysis of GITR expression on T cells, and is also commonly used in vitro as an agonistic antibody to induce GITR signaling in various assays.