Anti-LY76 Rat Antibody (violetFluor® 450) [clone: TER-119]
Catalog # 75-5921-U100
Supplier: Tonbo Biosciences
Specifications
- Antigen name:Lymphocyte Antigen 76
- Antigen symbol:LY76
- Clone:TER-119
- Conjugation:VioletFluor® 450
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Gene ID:104231
- Antigen synonyms:Ter119|TER-119
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Flow Cytometry
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The TER-119 antibody is named for the antigen to which it binds, a 52 kDa surface protein that is associated with glycophorin-A. TER-119 is considered to be a lineage marker for later stages of erythroid cell development, as its expression begins at the pro-erythroblast stage. TER-119 antigen is not expressed at either BFU-E or CFU-E stages, i.e. prior to the pro-erythroblast stage.