Anti-LEDGF Rabbit polyclonal antibody
Catalog # NOVUNB110-40561
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:LEDGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Gene ID:11168
- Antigen synonyms:PC4 and SFRS1 interacting protein 1|Dense fine speckles 70 kDa protein|p52|transcriptional coactivator p52/p75|PC4 and SFRS1-interacting protein|p75|Lens epithelium-derived growth factor|Transcriptional coactivator p75/p52|PSIP2|DFS70|MGC74712|PAIP|CLL-associated antigen KW-7|LEDGFDFS 70|PC4 and SFRS1 interacting protein 2
- Storage buffer:TBS and 0.1% BSA
- Storage temperature:Store at 4 °C. Do not freeze.
- Concentration:0,2 mg/ml
- Immunogen:The immunogen recognized by this antibody maps to a region between 480 and the C-terminus(residue 530) of human Lens Epithelium Derived Growth Factor/Transcriptional Coactivator p75 using the numbering given in entry NP_150091.2 (GeneID 11168).
- Purification:Immunogen affinity purified
- Pk:0,1 mL
Specifications
About this item
Rabbit Polyclonal LEDGF Antibody. Tested Applications: Western Blot, Immunoprecipitation. Tested Reactivity: Human.
Type: Primary
Antigen: LEDGF
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human