Anti-CD277/BTN3A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BT3.1]
Catalog # NOVUNBP2-00227PE
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:CD277/BTN3A1
- Clonality:Monoclonal
- Clone:BT3.1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Gene ID:11119
- Storage buffer:PBS
- Storage temperature:Store at 4 °C in the dark.
- Immunogen:The immunogen for this antibody is CD277 (B7 family, Butyrophilin).
- Purification:Protein A or G purified
- Size:100 μl
- Pk:0,1 mL
Specifications
About this item
The CD277 / BTN3A1 Antibody (BT3.1) [PE] from Novus Biologicals is a mouse monoclonal antibody to CD277 / BTN3A1. This antibody reacts with human. The CD277 / BTN3A1 Antibody (BT3.1) [PE] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: CD277/BTN3A1
Clonality: Monoclonal
Clone: BT3.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human