Anti-BARHL1 Rabbit Polyclonal Antibody
NOVUNBP1-86513
- Antibody type:Primary
- Antigen symbol:BARHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Gene ID:56751
- Antigen synonyms:BarH (Drosophila)-like 1|BarH-like homeobox 1|barH-like 1 homeobox protein
- Storage buffer:PBS (pH 7,2) and 40% Glycerol
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LELSPRSESSSDCSSPASPGRDCLETGTPRPGGASGPGLDSHLQPGQLSAPAQSRTVTSSFLIRDILADCKPLAACAPYSSSGQPAAPEPGGRLAAKAAEDFRDKLDKSGSNASSDSEYKVKEEGDREISSSRDSP
- Purification:Immunogen affinity purified
- Size:100 μl
- Pk:0,1 mL
The BARHL1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to BARHL1. This antibody reacts with human. The BARHL1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: BARHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human