Anti-TFF3 Rabbit Polyclonal Antibody
Catalog # NOVUNBP1-76514
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Trefoil Factor 3
- Antigen symbol:TFF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:7033
- Antigen synonyms:trefoil factor 3 (intestinal)|HITF|human intestinal trefoil factor10Intestinal trefoil factor
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human TFF3. The immunogen is located within the last 50 amino acids of TFF3.
- Purification:Immunogen affinity purified
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
The TFF3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TFF3. This antibody reacts with human, mouse, rat. The TFF3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TFF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat