Order Entry
Norway
ContactUsLinkComponent
 
Anti-PACAP/ADCYAP1 Goat Polyclonal Antibody
  NOVUNBP1-51959
undefined
Anti-PACAP/ADCYAP1 Goat Polyclonal Antibody
  NOVUNBP1-51959

 

  • Antibody type:
    Primary
  • Antigen name:
    Adenylate Cyclase activating Polypeptide 1
  • Antigen symbol:
    PACAP/ADCYAP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    116
  • Antigen synonyms:
    pituitary adenylate cyclase-activating polypeptide|PACAPMGC126852|adenylate cyclase activating polypeptide 1 (pituitary)
  • Storage buffer:
    0,5 mg/ml Tris (pH 7,3) and 0,5% BSA
  • Storage temperature:
    Store at -20 °C. Avoid freeze-thaw cycles.
  • Concentration:
    0,5 mg/ml
  • Immunogen:
    Peptide with sequence C-GKRYKQRVKNKGRR corresponding to C-Terminus according to NP_001108.2.
  • Tested applications:
    Peptide
  • Purification:
    Immunogen affinity purified
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

The PACAP / ADCYAP1 Antibody from Novus Biologicals is a goat polyclonal antibody to PACAP / ADCYAP1. This antibody reacts with human. The PACAP / ADCYAP1 Antibody has been validated for the following applications: Western Blot, Peptide ELISA.

Type: Primary
Antigen: PACAP/ADCYAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human