Order Entry
Norway
ContactUsLinkComponent
Membrane protein placenta tissue lysate (adult normal)
Membrane protein placenta tissue lysate (adult normal)
Catalog # NOVUNB820-60607
Supplier:  Novus Biologicals
CAS Number:  
Membrane protein placenta tissue lysate (adult normal)
Catalog # NOVUNB820-60607
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Cell Format:
    Cryopreserved
  • Pk:
    0,1 mg
  • Tested applications:
    WB, ImmunoPrecipitation

Specifications

About this item

Membrane protein placenta tissue lysate (adult normal).

Tested applications: Western blot and immunoprecipitation

Tested reactivity: Human