ISO safety signs - Fire alarm flashing light
Supplier: Brady
Certificates
About this item
When working in a loud production environment, ear protection is necessary to reduce noise exposure. But can employees still hear the fire alarm?
- Increase safety using internationally recognised signs
- Compliant with ISO 7010/ISO 3864
- Designs available in multiple top selling materials
To solve this issue, fire alarm flashing lights have been developed. ISO recognised the need to clearly indicate the location of fire alarm flashing lights. Production areas usually have many flashing lights which could also warn about the active status of a machine, or exposed moving components. To clearly distinguish the fire alarm from all other lights, ISO developed new signs that enable employees to quickly recognise they need to evacuate.
Certifications: ISO 7010/ISO 3864.