Sodium acetate, Corning® 3M™
MDTC46-033-CI
: Corning
:
- Pk:100 mL
- Description:3M™ sodium acetate, 100 ml
- Storage temperature:Ambient
- Shipping temperature:Ambient
Sodium Acetate is a common molecular biology reagent. All production lots are tested to ensure that they are free of RNAse. It is often used in buffer systems to maintain the pH of a given solution.