Order Entry
Norway
ContactUsLinkComponent
Carbenicillin disodium salt, powder
Carbenicillin disodium salt, powder
Catalog # MDTC46-100-RG
Supplier:  Corning
CAS Number:  
undefined
Carbenicillin disodium salt, powder
Catalog # MDTC46-100-RG
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :422.363 g/mol
Storage Temperature:Refrigerator
CAS Number:4800-94-6
Formula:C₁₇H₁₆N₂O₆S·2Na
MDL Number:MFCD00077683