Accessories for laboratory balances verified, France country-specific, Secura®
Catalog # SARBYSH30
Supplier: Sartorius Balances
Specifications
- Description:Filter holder (150 mm) enables flawless weighing of filters up to a diameter of 150 mm, designed for use in semi-micro and analytical balances
- Pk:1