Cleanroom gloves, nitrile, non sterile, light blue, tacky finish, PUREZERO® HG3
Supplier: Halyard
Certificates
About this item
High quality gloves designed to exact standards to meet the requirements of a contamination controlled environment.
- High quality nitrile
- Non sterile
- Accelerator-free formula reduces risk of type IV allergies
- For use in cleanroom environments (ISO 5/6/7/8, grade B/C/D aseptic)
- REACH Annex XVII
Offering the right glove for the right controlled environment, Halyard PUREZERO® provides a cleanroom glove with guaranteed low particulate counts and endotoxin levels due to repeated washes in deionised water and chlorination. Nickel-, powder-, silicon- and latex-free, with no phthalates. These gloves are formulated with zero presence of these hazardous traditional accelerators: Sulphur, thiurams, thiazoles, guanidine and carbamates (ZDEC or ZDBC). All backed up by a vertically integrated supply chain.
Certifications: These cleanroom gloves are certified as PPE category III per the Regulation (EU) 2016/425. The applicable standards are:
• EN ISO 374-1:2016 Type C K - dangerous chemicals and microorganisms (permeation time)
• EN ISO 374-2:2019 - dangerous chemicals and microorganisms (determining resistance to penetration holes/defects)
• EN ISO 374-4:2019 - resistance to degradation by chemicals
• EN ISO 374-5:2016 - microorganism and virus protection
• EN 420:2003 + A1:2009 - general requirements for protective gloves
• EN 16523-1:2015 + A1:2018 - permeation of chemicals (+IPA 70%**)
• ISO 16604:2004 - contact with blood and body fluids
• ASTM D6978 - permeation testing on chemotherapy drugs (14 drugs)
• EN ISO 374-1:2016 Type C K - dangerous chemicals and microorganisms (permeation time)
• EN ISO 374-2:2019 - dangerous chemicals and microorganisms (determining resistance to penetration holes/defects)
• EN ISO 374-4:2019 - resistance to degradation by chemicals
• EN ISO 374-5:2016 - microorganism and virus protection
• EN 420:2003 + A1:2009 - general requirements for protective gloves
• EN 16523-1:2015 + A1:2018 - permeation of chemicals (+IPA 70%**)
• ISO 16604:2004 - contact with blood and body fluids
• ASTM D6978 - permeation testing on chemotherapy drugs (14 drugs)
Packaging: Gloves are double-bagged, per 100 pieces.
Specifications
- Catalog No:
- 112-6532
- 112-6535
- 112-6536
- 112-6533
- 112-6534
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Tacky, with textured fingertips
- Tacky, with textured fingertips
- Tacky, with textured fingertips
- Tacky, with textured fingertips
- Tacky, with textured fingertips
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Finger thickness:
- 0,1 mm
- 0,1 mm
- 0,1 mm
- 0,1 mm
- 0,1 mm
- Palm thickness:
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- Anti-static:
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +