Terrestrial (Cypripedium) orchid medium, contains 600 mg/L calcium nitrate
Supplier: Apollo Scientific
Certificates
About this item
Contains 600 mg/L calcium nitrate and 200 mg/L casein; without ammonium nitrate.
- Plant tissue culture tested
Contains 600 mg/L calcium nitrate and 200 mg/L casein; without ammonium nitrate.