Order Entry
Norway
ContactUsLinkComponent
Gamborg's B-5 basal salt medium
Gamborg's B-5 basal salt medium
Catalog # APOSPMG398A-10X1L
Supplier:  Apollo Scientific
CAS Number:  
Gamborg's B-5 basal salt medium
Catalog # APOSPMG398A-10X1L
Supplier:  Apollo Scientific
CAS Number:  
Specifications
  • Pk:
    10
  • Description:
    Gamborg's B-5 basal salt medium 10×1 L
  • Seq:
    01
Specifications
About this item
Contains macro/micronutrients as described by Gamborget al. 1968. Plant tissue culture tested. Full specification given in appendix.
  • MFCD00240253