H1 oat medium
Supplier: Apollo Scientific
Certificates
About this item
Contains agar and sucrose. Contains the macro- and micronutrients and supplements required for the symbiotic culture of orchids.
- Plant tissue culture tested
Contains agar and sucrose. Contains the macro- and micronutrients and supplements required for the symbiotic culture of orchids.