Order Entry
Norway
ContactUsLinkComponent
Human recombinant PDGF-AA (from Yeast)
Human recombinant PDGF-AA (from Yeast)
Supplier:  Apollo Scientific
Certificates
Human recombinant PDGF-AA (from Yeast)
Supplier:  Apollo Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Yeast
  • Species:
    Human
  • Protein synonyms:
    Platelet Derived Growth Factor-AA
  • Protein/peptide name:
    PDGF-AA

Specifications

Frequently Bought Together