Recombinant Tick-Borne Encephalitis Virus Core
Catalog # APOSBITP1926-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Protein synonyms:Encephalitis Core
- Protein/peptide name:Tick-Borne Encephalitis Virus Core