Recombinant Hepatitis C Virus Nucleocapsid (core) 22kDa, Rhodamine
Supplier: Apollo Scientific
Certificates
About this item
Specifications
- Conjugation:Rhodamine
- Protein/peptide type:Recombinant
- Protein synonyms:HCV 22kDa
- Protein/peptide name:Hepatitis C Virus Nucleocapsid (core) 22kDa
Specifications
Frequently Bought Together