Order Entry
Norway
ContactUsLinkComponent
Mouse recombinant VEGF (from Sf9 cells)
Mouse recombinant VEGF (from Sf9 cells)
Supplier:  Apollo Scientific
Certificates
Mouse recombinant VEGF (from Sf9 cells)
Supplier:  Apollo Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells (Sf9)
  • Species:
    Mouse
  • Protein synonyms:
    Vascular Endothelial Growth Factor
  • Protein/peptide name:
    VEGF

Specifications

Frequently Bought Together