Human recombinant SDF-1a
Catalog # APOSBITC2044-10UG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Species:Human
- Seq:05
- Protein synonyms:Stromal Cell-Derived Factor-1 alpha, CXCL12
- Protein/peptide name:SDF-1a