NANOCOLOR® Photometric Tube Test Kit for Nickel
MANA985071
:
- Kjemisk metode:Dimethylglyoxime
- Fp:1 KIT
- Beskrivelse:NANOCOLOR® Nickel 4, test no. 0-71
- Dimensjoner:115×161×124 mm (including packaging)
- Modell:MANA985071
- Platform:NANOCOLOR® tube tests
- Lagringstemp:15…25 °C
- Holdbarhetstid (måneder):24
- Vekt:335 g including packaging
- Til enhet:UV/VIS II, VIS II, 500 D, PF-12Plus, Advance, MT Easy VIS, MT Easy UV
- Tester per kit:20
- Område:0,10 - 7,00 mg/L (Ni²⁺)
Photometric test kit for the determination of nickel in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers Precisely pre-dosed reagents with constant high quality from batch to batch Coloured pictogram instructions and easy handling of the tubes 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Sea water analysis: Yes Hazardous material: Yes Remark: Measuring range on NANOCOLOR VIS II
: Supplied in rugged box with 20 test tubes.