Gå til Overskrift
Gå til Hovedinnhold
Gå til Bunntekst
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-2,3-difluorobenzoic acid
4-Bromo-2,3-difluorobenzoic acid 98%
4-Bromo-2,3-difluorobenzoic acid 98%
:
Apollo Scientific
4-Bromo-2,3-difluorobenzoic acid 98%
:
Apollo Scientific
:
194804-91-6
:
:
C₇H₃BrF₂O₂
:
MFCD07368121
:
236.9983 g/mol
:
195…196 °C
:
GHS07
:
Kjøleskap