Order Entry
Norway
ContactUsLinkComponent
1,3-Dichloro-7-fluoroisoquinoline 98%
1,3-Dichloro-7-fluoroisoquinoline 98%
Catalog # APOSPC7422-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Dichloro-7-fluoroisoquinoline 98%
Catalog # APOSPC7422-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.0392 g/mol
Storage Temperature:Ambient
CAS Number:941294-25-3
Formula:C₉H₄Cl₂FN
MDL Number:MFCD09763685