kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline
Irriterende
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline
Leverandør:
Apollo Scientific
Sertifikat
Expand image
Irriterende
4-Chloro-8-fluoro-2-(trifluoromethyl)quinoline
Leverandør:
Apollo Scientific
Om dette produktet
CAS-nummer:
401567-85-9
:
Formel:
C₁₀H₄ClF₄N
MDL-nummer:
MFCD01570536
:
249.5921 g/mol
:
GHS07
Lagringstemperatur:
Ambient