Order Entry
Norway
ContactUsLinkComponent
2,3-Difluoro-4-ethoxyphenylboronic acid 98%
Irritant
2,3-Difluoro-4-ethoxyphenylboronic acid 98%
Catalog # APOSPC7277-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,3-Difluoro-4-ethoxyphenylboronic acid 98%
Catalog # APOSPC7277-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :201.96 g/mol
Melting Pt:151…154 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:212386-71-5
Formula:C₈H₉BF₂O₃
MDL Number:MFCD09258743